Gallic acid, anhydrous

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 3,4,5-Trihydroxybenzoic acid

G1034-100GM G1034-2.5KG G1034-500GM
75872-412EA 132.39 USD
75872-412 75872-414 75872-416
Gallic acid, anhydrous
Gallic acid

Gallic Acid, Anhydrous, also known as 3,4,5-trihydroxybenzoic acid, is a type of phenolic acid that is found in sumac, gallnuts and tea leaves, and is used in the pharmaceutical industry.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₆O₅
MW: 170.12 g/mol
Boiling Pt: 501
Storage Temperature: Ambient
MDL Number: MFCD00002510
CAS Number: 149-91-7
Merck Index: 13,04366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR