N-(3-Trifluoromethylphenyl)anthranilic acid

Supplier: Spectrum Chemical Mfg. Corp.

F3020-25GM F3020-5GM
75872-514EA 151.95 USD
75872-514 75872-516
N-(3-Trifluoromethylphenyl)anthranilic acid
N-(3-Trifluoromethylphenyl)anthranilic acid
Formula: C₁₄H₁₀F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00002422
CAS Number: 530-78-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR