3,4-Dimethoxyaniline

Supplier: Spectrum Chemical Mfg. Corp.

D1558-100GM D1558-25GM
75872-290EA 411.05 USD
75872-290 75872-292
3,4-Dimethoxyaniline
3,4-Dimethoxyaniline
Formula: C₈H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00008394
CAS Number: 6315-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR