Chlorprothixene hydrochloride
Supplier: Spectrum Chemical Mfg. Corp.
C3976-200GMBL
C3976-25GM
75872-188EA
75.85
USD
75872-188
75872-190
75872-192
Chlorprothixene hydrochloride
Chlorprothixene hydrochloride
Formula:
C₁₈H₁₈ClNS·HCl Storage Temperature: Refrigerator |
MDL Number:
MFCD01941605 CAS Number: 6469-93-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...