trans-1,4-Cyclohexanedicarboxylic acid

Supplier: Spectrum Chemical Mfg. Corp.

C1685-100GM C1685-25GM
75872-124EA 586.15 USD
75872-124 75872-126
trans-1,4-Cyclohexanedicarboxylic acid
trans-1,4-Cyclohexanedicarboxylic acid
Formula: C₈H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00066203
CAS Number: 619-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR