L(+)-Arginine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH

AR120-100GM AR120-1KG AR120-25GM AR120-12KGBL AR120-50KGBL
75871-892EA 115.58 USD
75871-892 75871-894 75871-896 76014-216 76014-218
L(+)-Arginine
L(+)-Arginine

L-Arginine, abbreviated as Arg, plays an important role in cell division, the healing of wounds, removing ammonia from the body, immune function, and the release of hormones.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR