Dimethyl acetal, Technical Grade

Supplier: Spectrum Chemical Mfg. Corp.

A2204-100ML A2204-25ML
75871-786EA 257.75 USD
75871-786 75871-788
Dimethyl acetal, Technical Grade
Dimethyl acetal
Formula: C₄H₁₀O₂
Density: 0.852
Storage Temperature: Ambient
MDL Number: MFCD00008493
CAS Number: 534-15-6
UN: 2377
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR