Citric acid, anhydrous 99.5-100.5% (alkalimetric, calculated on anhydrous substance), fine-granular, EMPROVE® ESSENTIAL USP, JP, E330, Ph. Eur., BP, FCC, SAFC®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

1.00247.5000 1.00247.9029
EM1.00247.5000CS 2160.95 USD
EM1.00247.5000 EM1.00247.9029
Citric acid, anhydrous 99.5-100.5% (alkalimetric, calculated on anhydrous substance), fine-granular, EMPROVE® ESSENTIAL USP, JP, E330, Ph. Eur., BP, FCC, SAFC®
Citric acid

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1 to 3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.


Conforms to the purity criteria on food additives according to the current European Commission Regulation.

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310 Dec.
Melting Pt: 153 °C Dec.
Density: 1.665 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Alkalimetric, Calculated on Anhydrous Substance) 99.5 - 100.5%
Identity (IR-Spectrum) Passes Test
Appearance White to Almost White, Fine-Granular Powder
Appearance of Solution (200 g/l; Water) Clear (≤3 NTU) and Not More Intense in Color than Reference Solution Y₇, BY₇ or GY₇
Sulfate (SO₄) ≤50 ppm
Heavy Metals (as Pb) ≤5 ppm
Al (Aluminium) ≤0.2 ppm
As (Arsenic) ≤1 ppm
Hg (Mercury) ≤1 ppm
Pb (Lead) ≤0.5 ppm
Oxalates (as C₂H₂O₄) ≤100 ppm
Residual Solvents (ICH (Q3C)) Excluded by Manufacturing Process
Readily Carbonisable Substance Passes Test
Sulfated Ash (600 °C) ≤0.05%
Water (According to Karl Fischer) ≤0.5
Bacterial Endotoxins 0.5
Corresponds to Ph. Eur., BP , JP, USP, E330, FCC


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR