Sodium metabisulfite, dry 97-100.5%, Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., E223, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite

13459-6X1KG 13459-4X5KG
BJ13459-1KGCS 288.6 USD
BJ13459-1KG BJ13459-5KG
Sodium metabisulfite, dry 97-100.5%, Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., E223, Fluka™
Sodium metabisulphite
Formula: Na₂S₂O₅
MW: 190.11 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt: >150 °C (dec.)
Density: 2.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97.0 - 100.5 %
Assay of Sulfur Dioxide 65.0 - 67.4 %
pH (5 %, 20°C) 3.5 - 5.0
pH (10% solution) 4.0 - 5.0
Arsenic (As) Max. 2 ppm
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 2 ppm
Selenium (Se) Max. 5 ppm
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 0.01 %
Thiosulfate (S₂O₃) complying
Appearance of the solution complying
Residual solvents complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR