Potassium chlorate ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

31247-25KG 31247-500G
BJ31247-100GEA 57.62 USD
BJ31247-100G BJ31247-25KG BJ31247-500G
Potassium chlorate ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Potassium chlorate
Formula: KClO₃
MW: 122.55 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 356 °C
Density: 2.34 g/cm³ (23 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011361
CAS Number: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II

Order Now

Specification Test Results

Assay Min. 99 %
Water insoluble matter Max. 0.005 %
Arsenic (As) Max. 0.00005 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 0.0003 %
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.01 %
Heavy metals (as Pb) Max. 0.0005 %
Bromate (BrO₃) Max. 0.015 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR