Lithium carbonate ≥99%, Purum, Fluka™

Supplier: Honeywell Research Chemicals
Warning

13010-100KG 13010-6X1KG
BJ13010-100GEA 97.42 USD
BJ13010-100G BJ13010-100KG BJ13010-1KG
Lithium carbonate ≥99%, Purum, Fluka™
Lithium carbonate
Formula: Li₂CO₃
MW: 73.89 g/mol
Boiling Pt: 1.300 °C
Melting Pt: 720
Density: 2.110 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Specification Test Results

Assay Min. 99 %
Calcium (Ca) Max. 0.03 %
Iron (Fe) Max. 0.003 %
Potassium (K) Max. 0.03 %
Magnesium (Mg) Max. 0.01 %
Sodium (Na) Max. 0.1 %
Heavy metals (as Pb) Max. 0.003 %
Chloride (Cl) Max. 0.02 %
Sulfate (SO₄) Max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR