Lead(II) nitrate ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Lead dinitrate

228621-2.5KG 228621-500G
BJ228621-100GEA 59.54 USD
BJ228621-100G BJ228621-2.5KG BJ228621-500G
Lead(II) nitrate ≥99.0% ACS, Fluka™
Lead (II) nitrate
Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 458…459 °C (1.023 hPa)
Density: 4.49 g/cm³ (20.1 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
In diluted HNO₃ insoluble matter Max. 0.005 %
Calcium (Ca) Max. 50 ppm
Copper (Cu) Max. 20 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Sodium (Na) Max. 200 ppm
Chloride (Cl) Max. 10 ppm
Form Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR