Diatomaceous earth, powder suitable for most filtrations, Fluka™

Supplier: Honeywell Research Chemicals
Warning

D3877-1KG D3877-500G D3877-5KG
BJD3877-1KGEA 176.14 USD
BJD3877-1KG BJD3877-500G BJD3877-5KG
Diatomaceous earth, powder suitable for most filtrations, Fluka™
Formula: SiO₂
Melting Pt: 450 °C
Density: 2.4 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 61790-53-2
EINECS: 293-303-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR