Diammonium hydrogen orthophosphate ≥98% (by alkalimetry), Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
30402-1KG
30402-500G
30402-50KG
BJ30402-1KGEA
226.08
USD
BJ30402-1KG
BJ30402-500G
BJ30402-50KG
Diammonium hydrogen orthophosphate ≥98% (by alkalimetry), Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
di-Ammonium hydrogen orthophosphate
Formula:
(NH₄)₂HPO₄ Melting Pt: 155 °C Density: 1.62 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 EINECS: 231-987-8 |
Specification Test Results
Assay | Min. 98.0 % |
Water insoluble matter | Max. 0.005 % |
pH (5 %, 25 °C) | 7.7 - 8.1 |
Calcium (Ca) | Max. 0.001 % |
Iron (Fe) | Max. 0.001 % |
Potassium (K) | Max. 0.005 % |
Magnesium (Mg) | Max. 0.0005 % |
Sodium (Na) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 0.001 % |
Chloride (Cl) | Max. 0.001 % |
Nitrate (NO₃) | Max. 0.003 % |
Sulfate (SO₄) | Max. 0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...