Stopcocks, High Vacuum, Single Oblique Bore, Hollow Glass Plugs, Chemglass
Supplier: Chemglass
CG-472-01 CG-472-02
80074-314EA 209.82 USD
80074-314 80074-316
Stopcocks, High Vacuum, Single Oblique Bore, Hollow Glass Plugs, Chemglass
Stopcocks
Hollow plug, oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated via a separate hole in the plug which is not in the line of flow.