di-Lithium tetraborate ≥99.9% (trace metals basis) PPA
Supplier: Honeywell Research Chemicals
Synonyms:
Lithium tetraborate, Dilithium tetraborate
222534-1KG
222534-250G
BJ222534-1KGEA
649.39
USD
BJ222534-1KG
BJ222534-250G
di-Lithium tetraborate ≥99.9% (trace metals basis) PPA
di-Lithium tetraborate
Formula:
Li₂B₄O₇ MW: 169.12 g/mol Melting Pt: 760…880 °C Density: 0.25 g/cm³ (25 °C) Storage Temperature: Room Temperature |
MDL Number:
MFCD00011083 CAS Number: 12007-60-2 EINECS: 234-514-3 |
Specification Test Results
Loss on ignition (800 °C, 1 h) | Max. 1.00 % |
Grain dissipation (medium value) | Max. 500 µm |
Metal trace analysis (ICP) | Max. 1000 ppm |
Boron (B) | Min. 25.0 % |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...