Glycerol-1,2-carbonate ≥90.0% (by GC)

Supplier: Spectrum Chemical Mfg. Corp.

G1488-100GM G1488-25GM
75872-534EA 332.02 USD
75872-534 75872-536
Glycerol-1,2-carbonate ≥90.0% (by GC)
Glycerol-1,2-carbonate
Formula: C₄H₆O₄
Storage Temperature: Ambient
MDL Number: MFCD00085561
CAS Number: 931-40-8

Order Now

Specification Test Results

Purity (GC) Min. 90.0%
Refractive Index (20 °C) 1.4570 - 1.4610


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR