Syringe Pump, CMA 4004, Harvard Apparatus
Supplier: Harvard Apparatus
CMA400400
MSPP-CMA400400EA 8623.08 USD
MSPP-CMA400400
Syringe Pump, CMA 4004, Harvard Apparatus
Pumps Syringe Pumps
CMA 4004 Syringe pump is easy-to-use, can hold four-syringes, and provides a very broad flow range rate suitable for micro dialysis. The pump can deliver very precise micro-injections that can be repeated in intervals.
  • Quick setup using the built-in syringe library
  • Program simple to complex methods without a PC
  • Email methods to your peers, download methods and upgrade software remotely
  • Set the start/stop time for the pump
High resolution LCD color touch screen makes it very easy to program and recall methods. The direction of flow can easily be reversed for experiments requiring fluid withdrawal.
Syringe pump has the ability to run in a horizontal or vertical orientation. This allows choosing the proper orientation based on the experiment setup. Equipped with RS-485 for daisy chaining pumps and Digital I/O for remote control as well as USB serial port for computer control.