Iron(III) chloride 0,0250% (w/v) in aqueous solution
Supplier: AQUA SOLUTIONS MS
Synonyms:
Iron trichloride, Ferric chloride
3780-500ML
3780-1L
101226-996EA
30.05
USD
101226-996
101223-038
Iron(III) chloride 0,0250% (w/v) in aqueous solution
Iron (III) chloride
Formula:
FeCl₃ |
MDL Number:
MFCD00011005 CAS Number: 7705-08-0 |
Specification Test Results
Prepared to formulation on file | Yes |
W/V% Ferric chloride | 0.0247 - 0.0253 |
Appearance and color | Clear orange liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...