Lead(II) acetate trihydrate 99.0-103.0% (by titrimetric analysis) ACS

Supplier: CHEM IMPEX INTL INC MS
Danger

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

22942-1KG 22942-2.5KG 22942-100G
102552-994EA 91.63 USD
102552-994 102559-360 102624-456
Lead(II) acetate trihydrate 99.0-103.0% (by titrimetric analysis) ACS
Lead (II) acetate trihydrate

Can be used in the detection of sulfide, chromate, and molybdenum trioxide.

Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Melting Pt: ~75 °C
Density: 2.550 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150023
CAS Number: 6080-56-4
UN: 1616
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Anion, Chloride (Cl⁻) ≤5 ppm
Anion, Nitrate and Nitrite (as NO₃) ≤0.005%
Cation, Calcium (Ca) ≤0.005%
Cation, Copper (Cu) ≤0.002%
Cation, Iron (Fe) ≤0.001%
Cation, Potassium (K) ≤0.005%
Cation, Sodium (Na) ≤0.01%
Insoluble Material ≤0.01%
Assay by Titration 99.0 - 103.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR