SMD Hot Air Rework Station, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FR810B-05 FR810B-05
76248-906EA 2963.14 USD
76248-906 76250-530
SMD Hot Air Rework Station, American Hakko Products
Soldering Supplies Soldering Stations
These stations are designed with vacuum pick up function for easy and safe removal of components.
  • Easy to read digital display for both temperature and air level
  • Slender hand piece with integrated start/stop button
Ability to store up to 5 user programmable presets for temperature, airflow and time.
Delivery information: Supplied with handpiece, nozzle (Φ 4 mm), handpiece holder, Vacuum pipe control knob L (with screw), pads (qty 2 each of Ø3 mm, Ø5 mm, Ø7.6 mm), heat resistant pad, power Cord, temperature distribution chart and instruction manual.