Acridine orange (hemizinc salt), redish brown powder biological stain
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride
00481-5G
00481-25G
00481-100G
00481-250G
102615-288EA
30.58
USD
102615-288
102615-290
102615-292
102615-294
Acridine orange (hemizinc salt), redish brown powder biological stain
Acridine orange (hemizinc salt)
Formula:
C₁₇H₂₀ClN₃·HCl·½ZnCl₂ MW: 369.96 g/mol Melting Pt: 285 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081043 CAS Number: 10127-02-3 |
Specification Test Results
Loss on Drying | ≤1% |
Dye Content | ≥90.0% |
Absorption at Lambda Max at 492 nm | ≥0.514 (5.0 mg/l in Water) |
Absorption at Lambda Max at 269 nm | ≥0.514 (5.0 mg/l in Water) |
Extinction Coefficient at 489 - 495 nm | ≥38000 |
Extinction Coefficient at 265 - 271 nm | ≥38000 |
Solubility | Clear, orange to brown solution (1 mg/mL in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...