Rad-Sequel™ Safety Eyewear, Radians
Supplier: RADIANS INC.(FKA TECHNA INTL) TE
RS1-B RS1-10
76243-254EA 6.19 USD
76243-254 76243-974
Rad-Sequel™ Safety Eyewear, Radians
Safety Glasses
Rad-Sequel™ safety glasses have a lightweight design which provides a comfortable fit.
  • Rubber tipped temples provide a secure fit
  • Soft, rubber nose piece provides all day comfort
Protective lens provide 99.9% UVA/UVB protection. Polycarbonate lens provides impact resistance.
Certifications: Safety Glasses meet ANSI Z87.1+ standards.