Reamers, Cleaveland, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
C25858 C25226
76250-092EA 151.09 USD
76250-092 76250-942
Reamers, Cleaveland, Cutting Tool Supply
Tool Sets
Reamers made of high speed steel.
  • With straight flute and shank
  • With thru holes
Certifications: Complies to ASME B94.2 and DIN 338.