Grinding Stones, Dremel Services
Supplier: DREMEL SERVICES INC. TE
DR-8153 DR-8193
76249-340EA 13.47 USD
76249-340 76249-342
Grinding Stones, Dremel Services
Tool Sets
A high-quality industrial abrasive made for extended general-purpose grinding on metals, castings, welded joints, rivets and rust.
  • Ideal for sharpening, deburring and general purpose grinding on most materials including stainless steel
  • Specially designed for working with conic surfaces