Solid Carbide Straight Shank Chucking Reamers, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
54098-MO 54094-MO 54090-MO
76249-132EA 90.65 USD
76249-132 76249-134 76249-136
Solid Carbide Straight Shank Chucking Reamers, Cutting Tool Supply
Tool Sets
Solid carbide reamers suitable for higher cutting speeds and longer tool life. High rigidity enhances hole accuracy and quality.
  • Resistant to wear and heat
Recommended for general reaming of ferrous and nonferrous materials including steel, alloy steel, stainless steel, plastic, aluminum and other abrasive non-ferrous materials.