Nitric acid 65% Huey, BAKER ANALYZED® for corrosion testing, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Nitric acid solution

Management of Change (MOC) category = NI

9597-33 9597-R
JT9597-33CS 659.58 USD
JT9597-33 JT9597-R
Nitric acid 65% Huey, BAKER ANALYZED® for corrosion testing, J.T.Baker®
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1),II
Merck Index: 13,06608

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as HNO₃) (by acidimetry) 64.8 - 65.2 %
Appearance Passes Test
Color (APHA) ≤ 10
Residue after Ignition ≤ 0.0005 %
Chloride (Cl) ≤ 0.05 ppm
Phosphate (PO₄) ≤ 0.20 ppm
Sulfate (SO₄) ≤ 0.5 ppm
Trace Impurities - Arsenic (As) ≤ 0.005 ppm
Trace Impurities - Chromium (Cr) ≤ 0.100 ppm
Trace Impurities - Copper (Cu) ≤ 0.050 ppm
Heavy Metals (as Pb) ≤ 0.1 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Nickel (Ni) ≤ 0.050 ppm
Fluoride (F) ≤ 1.000 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR