Clamp Gaskets, EDPM, Garlock Hygienic Technologies
Supplier: Rubber Fab
40MPE-100 40MPE-400
76238-126EA 1.76 USD
76238-126 76238-128
Clamp Gaskets, EDPM, Garlock Hygienic Technologies
Seals Gaskets
Tri-clamp sanitary gaskets for use in pharmaceutical industries as well as in Food and Beverage.
  • Blue gasket is X-ray detectable
  • For use with 1 inch (black) or 4 inch (blue) tubes
Tri-clamp sanitary gaskets fit into the grooves of a tri-clamp tube fitting. Tri-clamp fittings, also called tri-clover fittings, clamp together two tubes in a process line and the gasket provides a sterile, leakproof seal.
Certifications: Meets 21CFR 177.2600; passed U.S.P. Class VI Cytotoxicity testing.