Inch Nut Drivers, Wiha Tools
Supplier: WIHA TOOLS TE
26531 26539 26547
76242-308EA 27.82 USD
76242-308 76242-310 76242-312
Inch Nut Drivers, Wiha Tools
Nutdrivers
These nut drivers are with precision handle.
  • Inch sizes
  • Blade CVM steel
  • Hard chrome finished