Anti-IFNgamma Armenian hamster Monoclonal Antibody [clone: H22]

Supplier: Bio X Cell
BE0312-100MG BE0312-5MG BE0312-25MG BE0312-1MG BE0312-50MG
103721-636EA 3920 USD
103721-636 103721-638 103721-640 103721-642 103721-644 76555-698 76555-896 76556-034 76556-196 76557-142
Anti-IFNgamma Armenian hamster Monoclonal Antibody [clone: H22]
Antibodies
InVivoMAb anti-mouse IFNγ, Clone: H22, Size: 100 mg, Isotype: Armenian Hamster IgG, Applications: in vivo IFNγ neutralization, in vitro IFNγ neutralization

Type: Primary
Antigen: IFNgamma
Clonality: Monoclonal
Clone: H22
Conjugation:
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR