Horse Plasma Sterile In Sodium Heparin, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D507-06-1000 D507-06-0100 D507-06-0050 D507-06-0500
75934-574EA 1054.18 USD
75934-574 75934-576 75934-578 75934-580
Horse Plasma Sterile In Sodium Heparin, Rockland Immunochemicals
Blood
Sterile Plasma in Anticoagulant
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
used as a supplement to cell culture media
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances andMore Product Information