Human Recombinant CCL13
Supplier: Rockland Immunochemical
Recombinant Human CCL13 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Monocyte Chemotactic Protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as IL-1 and TNFα. MCP-4 is a ligand for three different G protein coupled receptors, CCR2, CCR3 and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils and basophils and this signaling is associated with the allergic response. Recombinant human MCP-4 is a non-glycosylated protein, containing 74 amino acids, with a molecular weight of 8.5 kDa.
Monocyte Chemotactic Protein-4 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Monocyte Chemotactic Protein-4 in immunological assays.