Human Recombinant CD40 Ligand
Supplier: Rockland Immunochemical
009-001-U68-0010 009-001-U68-0050
10800-688EA 2179 USD
10800-688 10800-692 10800-700
Human Recombinant CD40 Ligand
Proteins and Peptides
Recombinant Human CD40 Ligand control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-α, and IL-2. Recombinant human CD40 Ligand is a non-glycosylated protein, containing 149 amino acids, with a molecular weight of 16.3 kDa.
CD40 Ligand Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-CD40 Ligand in immunological assays.