Laptop Charging Carts, Luxor
Supplier: Luxor
LLTP12-B LLTP18-B LLTP24-B
76203-842EA 2046.44 USD
76203-842 76203-844 76203-846
Laptop Charging Carts, Luxor
Carts
Laptop charging cart is an ideal solution for charging and securing laptops or tablets.
  • Cabinet and shelves come fully assembled
  • All-steel construction and padded top surface
  • Rubber-coated dividers to organize and protect the tablet
  • Ample ventilation allows air to circulate freely through the unit
  • Each shelf holds nine laptops safely between padded dividers
  • Side cord wrap
The door locking system keeps the laptops secure while they are charging from the rear-mounted electrical strips. The lock comes with two sets of keys.
Holes in the base of the cart allow passage of the two power-strip cables. Rubber casters (4") ensure smooth transport from place to place. Two of the casters are lockable to secure the cart in place.
Certifications: Meets ANSI/BIFMA standards.