Phosphatase Inhibitor Cocktail IV, EMD Millipore
Supplier: MilliporeSigma
524628-1ML
524628-1SET
EM524628-1MLEA
183.12
USD
EM524628-1ML
EM524628-1SET
Phosphatase Inhibitor Cocktail IV, EMD Millipore
Cell Culture Supplements and Additives
Phosphatase inhibitor cocktail IV is a cocktail of three phosphatase inhibitors for the inhibition of both serine/threonine and alkaline phosphatases.
- Form: Liquid
- Formulation: In 95% DMSO
- Highly toxic
Caution: Irritating to eyes, respiratory system and skin. For long-term storage, aliquot and freeze (–20 °C). Dilute 1:50 just prior to use.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...