VWR® Zelite™ Tubing, PE
Supplier: NewAge Industries
251-0571-500 251-0256-500
76240-132EA 202.43 USD
76240-132 76240-134
VWR® Zelite™ Tubing, PE
Tubing Transfer Tubing
These tubing are made from non-toxic ingredients and does not impart a taste or odor to critical streams.
  • Flexible, lightweight, durable
  • Translucent natural or opaque colors
  • Made from 100% virgin grade raw materials
  • Good dielectric properties
Offers enhanced resistance to stress cracking and is chemically inert forming a good barrier to gases, vapors, and moisture.
Certifications: ZELITE conforms to the applicable regulations of the FDA for use of articles intended for contact with food.