Stainless Steel Tweezers, Lindstrom Precision Tools
Supplier: J.H. Williams Tool Group (Lindstrom)
2AX-SA TL-7A-SA LIND-TL-AA-SA
76238-878EA 98.71 USD
76238-878 76238-926 76239-796 76239-996 76239-998 76240-090 76240-092 76240-094
Stainless Steel Tweezers, Lindstrom Precision Tools
Forceps
Lindström tweezers offer perfect balance, tip alignment, and symmetry. The non-magnetic stainless steel provides excellent resistance to many acids because of the high concentration of molybdenum.
  • Suitable for continuous high temperature use
  • Anti-magnetic
  • Anti-acid
High precision reverse action: These are tweezers especially intended for applications where a high precise self-closing action is required (e.g. microscopy). They permit holding parts without finger pressure.
High precision: Primarily suitable for delicate standard applications and precision work.
Boley: Popular tweezers for general purpose applications.
General purpose: Serrated tips and handles for a secure grip.
Component handling: Specifically designed tips hold delicate electronic components firmly and safely. These tweezers can handle many types of round components and electronic devices such as resistors and diodes.