Adhesive Transfer Tapes, 3M
Supplier: 3M
7000123358 7100089859 7010372176
76242-690CS 1866.2 USD
76242-690 76242-692 76246-058
Adhesive Transfer Tapes, 3M
Tapes
Adhesive transfer tape provides outstanding adhesion on metal and high surface energy plastics. High performance acrylic adhesive 200MP delivers excellent shear strength to resist slippage and edge lifting. Temporarily repositionable for placement accuracy, this tape also performs well after exposure to humidity and hot/cold temperatures.
  • Delivers exceptional peel and shear strength for metals and HSE plastics
  • 58# polycoated kraft paper (PCK) liner offers a high strength bond for splicing and laminating applications
  • Thicker adhesives improve wet-out and adhesion on textured surfaces
  • Provides very high resistance to solvents and humidity
It offers excellent moisture and solvent resistance which enable it to form resilient, durable bonds that conform to a wide variety of applications.