The Ionized Air Gun provides efficient static charge neutralization by effectively removing particle contamination, even on some of the most static-sensitive products.
- Neutralize electrostatic charges to eliminate ElectroStatic Attraction (ESA), visual imperfections, and contamination by dislodging charged dust and debris
- Fast <1 second discharge times with ±30 volt offset voltage balance
- Use filtered air or nitrogen (no adjustment required); allows for quick change between compressed gas supply
- Console mounting hardware included for placement in the most convenient and productive locations
- Quick filter replacement feature minimizes down time
Modular design with 7' ESD protective hose with trigger switch allows operators freedom of motion and minimizes awkward or unsupported positions. The unit does not include a power adapter, and it must be purchased separately. NIST calibrated with certificate included. Calibrated with accepted procedures and standards traceable to the National Institute of Standards and Technology.
Certifications: Tested Per ANSI/ESD - STM 3.1 and ESD TR53. Meets ANSI/ESD S20.20 ionizer required limits. cULus, CE, RoHS2.
Ordering information: Power adapter sold separately.
Packaging: 100 bags per pack.