Retention Testers, Testequity LLC TE
Supplier: TESTEQUITY LLC TE
HT250-6 HT250-4
76247-344EA 293.52 USD
76247-344 76247-650
Retention Testers, Testequity LLC TE
Tool Sets
Ths testers to test retention of socket contacts, use the socket tester tip with a preset tool insert tester (pin) into mating end of contact.
Tool must be in a straight line with contact. For pin contacts, fit pin tester to tool body and place over contact at the mallng end.