Tape, PTFE Film, 3M
Supplier: 3M
7000132176 7010395800 7000132175 7000050115 7010380281
76242-376CS 6396.79 USD
76242-376 76242-696 76242-698 76242-700 76242-702 76242-704 76242-706 76242-710 76242-712 76245-018 76251-810
Tape, PTFE Film, 3M
Tapes
These tapes are used for heated machinery and equipment.
  • Get a high strength and abrasion resistance
  • Available in a wide selection of widths for custom processes
It uses an extruded polytetrafluoroethylene (PTFE) film backing, which has an extremely low coefficient of friction, in combination with a pressure sensitive silicone adhesive to create smooth, non-stick surfaces over a variety of substrates.
Its thicker backing helps extend the service life over thinner versions when subjected to abrasion and wear, and has a high strength and abrasion resistance that ensures reliability and performance to reduce friction, rattle and machinery noise.