TRIlites® 984 CP Tri-Polymer Gloves, Mapa Professionnel AdvanTech
Supplier: MAPA
34984027 34984028
89068-064CS 622.8 USD
89068-064 89068-066 89068-068 89068-070
TRIlites® 984 CP Tri-Polymer Gloves, Mapa Professionnel AdvanTech
Gloves
Unique tri-polymer blend of natural rubber, neoprene, and nitrile provides excellent product protection and optimum tactile sensitivity.
  • Comfort of latex with added strength, tear resistance, and chemical splash protection of nitrile and neoprene
  • Textured fingertips deliver superior grip in both wet and dry applications
  • Extra long, 11.5" length offers extended protection for the wrist and upper forearm
  • Powder-free process eliminates cross-contamination
  • Ambidextrous
Gloves are great for laboratory research, light assembly and sorting, precision partsMore Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Disposable/Reusable Disposable
Powdered/Powder-free Powder-Free
Cuff Style Beaded