2-Bromethylamine hydrobromide 98-101% (by titrimetric analysis)
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide
14308-250G
14308-1KG
14308-25G
14308-100G
14308-25G
102594-364EA
117.97
USD
102594-364
102594-366
102621-456
102621-458
CH14308-25G
2-Bromethylamine hydrobromide 98-101% (by titrimetric analysis)
2-Bromethylamine hydrobromide
Formula:
BrCH₂CH₂NH₃Br MW: 204.89 g/mol Melting Pt: 170…175 °C Storage Temperature: Freezer |
MDL Number:
MFCD00012886 CAS Number: 2576-47-8 |
Specification Test Results
Appearance | White to off-white crystalline powder |
Solubility | Colorless solution (50 mg/ml in water) |
Melting Point | 170 - 175 °C |
Water Content (Karl Fisher) | ≤0.5% |
pH | 3 - 5 |
Infrared Spectrum | Conforms to Structure |
Assay by titration | 98 - 101% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...