LB Agar Medium Large Capsules, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
- Proprietary media for enhanced cell yield
- Convenient packaging formats for a variety of applications and budgets
- Proven and reliable reputation
- High-quality raw materials and additives
- Encapsulated Media
- No weighing, dust and cleanup
- Drop capsules into water and autoclave
LB agar medium is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. The ultimate in ease and convenience, LB Agar Medium in a capsule form eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder.The growth characteristics are the same as LB agar powder. LB agar medium supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesize. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.
Certifications: Molecular Biology Certified™. Gelatin capsules are produced in a facility that meets the FDA’s recent guidance as being derived from BSE-free herds that are born, raised, and slaughtered in the United States.