Nitric acid ACS

Supplier: Ricca Chemical
Danger

Synonyms: Nitric acid solution

RABN0010-1C RABN0010-2.5D1 RABN0010-500D1
RCRABN0010-1CEA 178 USD
RCRABN0010-1C RCRABN0012.5D1 RCRABN001500D1
Nitric acid ACS
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8 (5.1),II

Order Now

Specification Test Results

Appearance Colorless to light yellow liquid
Assay 68.00-70.00 %
Color (APHA) max 10
Heavy Metals (as Pb) max 0.2 ppm
Sulfate (SO₄) max 1 ppm
Chloride (Cl) max 0.5 ppm
Iron (Fe) max 0.2 ppm
Arsenic (As) max 0.01 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR