Silicon(II) oxide on Cu grid, support films 15 – 30 nm for electron microscopy
Supplier: Electron Microscopy Sciences
Synonyms:
Silicium (II) oxide, Silicon monoxide, Silicon oxide
Support Film on grids has become a main product line for us since the demand for high quality coated grids has increased. To make your microscopy work easier and to save you a great deal of time we offer you a complete line. All of our coated grids are optically checked followed by batch testing in the EM.
A thin film of pure silicone Monoxide (15 – 30 nm) is deposited directly on top of the grid. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. All of our film is laid on the shiny side of the grid.
Formula:
SiO MW: 44.08 g/mol Boiling Pt: 1880 Melting Pt: 1702 |
MDL Number:
MFCD00151536 CAS Number: 10097-28-6 Merck Index: 12,08640 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...