Anti-IgG Fc Mouse Monoclonal Antibody [clone: N3318]

Supplier: TCI
M2977-1VIAL
TCM2977-VIALEA 204.97 USD
TCM2977-VIAL
Anti-IgG Fc Mouse Monoclonal Antibody [clone: N3318]
Antibodies
The monoclonal antibodies recognize the Fc region of human IgG.

This antibody reacts with human IgG on ELISA (1:5000) and Western blotting (1:1000).

Type: Primary
Antigen: IgG Fc
Clonality: Monoclonal
Clone: N3318
Conjugation: Unconjugated
Epitope: F(c)
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR