Ruthenium 5% on alumina powder, dry 70 µm for use as a catalyst, reduced , BASF Catalyst
Supplier: SPECTRUM CHEMICAL MFG CORP
SP-401-25G
SP-401-5G
SP-401-100G
76181-216EA
239.25
USD
76181-216
76181-218
76181-248
Ruthenium 5% on alumina powder, dry 70 µm for use as a catalyst, reduced , BASF Catalyst
Ruthenium
5 Percent Ruthenium on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst
Formula:
Ru MW: 101.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...