Platinum 5% on activated carbon powder wetted with water 27 µm, unreduced , BASF Catalyst

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Platinium

SP-202-100G SP-202-25G SP-202-5G
76124-452EA 1232.5 USD
76124-452 76124-454 76124-456
Platinum 5% on activated carbon powder wetted with water 27 µm, unreduced , BASF Catalyst
Platinum

5 Percent Platinum on Activated Carbon Powder, Unreduced, Water Wet, 27 microns

Formula: Pt
MW: 195.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR