Anti-MIP-1alpha (CCL3) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P77BT-25UG 500-P77BT-50UG
10775-708EA 339.8 USD
10775-708 10775-710 10775-712
Anti-MIP-1alpha (CCL3) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat MIP-1α (CCL3). Anti­Rat MIP-1α (CCL3)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: MIP-1alpha (CCL3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR